accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONOOCH3CH3

CAS RN 38428-14-7

IUPAC Name: 2-methylpropyl 2-(2-methylpropoxy)-1,2-dihydroquinoline-1-carboxylate
Synonyms: 2-methylpropyl 2-(2-methylpropoxy)-2H-quinoline-1-carboxylate
Molecular Weight (g/mol): 303.4
Molecular Formula: C18H25NO3
InChi Key: LPBHYOYZZIFCQT-UHFFFAOYNA-N
SMILES: CC(C)COC1C=CC2=CC=CC=C2N1C(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results