accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2(S)H2NHClHClCH3

CAS RN 3844-53-9

IUPAC Name: ethyl (2S)-2,6-diaminohexanoate dihydrochloride
Synonyms: ethyl (2S)-2,6-diaminohexanoate dihydrochloride
Molecular Weight (g/mol): 247.16
Molecular Formula: C8H20Cl2N2O2
InChi Key: DZIYAIZKJOHVQC-KLXURFKVSA-N
SMILES: Cl.Cl.CCOC(=O)[C@@H](N)CCCCN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results