accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OOCH3NH2HCl

CAS RN 3844-54-0

IUPAC Name: methyl (2S)-2-aminohexanoate hydrochloride
Synonyms: O-methyl-L-norleucine hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: FMMOVZRXLNVNBI-HQFIOVGONA-N
SMILES: Cl.CCCC[C@H](N)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)