accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38444-87-0

IUPAC Name: 3,3',5-trichloro-1,1'-biphenyl
Synonyms: 3,3',5-trichlorobiphenyl
Molecular Weight (g/mol): 257.54
Molecular Formula: C12H7Cl3
InChi Key: RIBGNAJQTOXRDK-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results