accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38465-55-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 629.54
Molecular Formula: C32H30N2NiS4
InChi Key: LDYCDKFAGIOUDE-JZONXAMZSA-J
SMILES: [Ni].CN(C)C1=CC=C(C=C1)C(\[S-])=C(\[S-])C1=CC=CC=C1.CN(C)C1=CC=C(C=C1)C(\[S-])=C(\[S-])C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results