accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 3848-24-6

IUPAC Name: hexane-2,3-dione
Synonyms: 2,3-hexanedione
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: MWVFCEVNXHTDNF-UHFFFAOYSA-N
SMILES: CCCC(=O)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)