accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CP

CAS RN 384842-24-4

IUPAC Name: dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphane
Synonyms: dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphane
Molecular Weight (g/mol): 390.55
Molecular Formula: C27H35P
InChi Key: YMSBPYCREGBACF-UHFFFAOYSA-N
SMILES: CC(P(C1CCCCC1)C1CCCCC1)=C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results