accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3852-11-7

IUPAC Name: methyl 3-methoxy-2-methylpropanoate
Synonyms: methyl 3-methoxy-2-methylpropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: LYLUAHKXJUQFDG-UHFFFAOYNA-N
SMILES: COCC(C)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results