accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOONH4

CAS RN 38542-94-8

IUPAC Name: ammonium trifluoromethanesulfonate
Synonyms: ammonium triflate
Molecular Weight (g/mol): 167.1
Molecular Formula: CH4F3NO3S
InChi Key: BMWDUGHMODRTLU-UHFFFAOYSA-N
SMILES: [NH4+].[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results