accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl2H2H2H2H

CAS RN 3855-82-1

IUPAC Name: dichloro(²H₄)benzene
Synonyms: dichloro(²H₄)benzene
Molecular Weight (g/mol): 151.02
Molecular Formula: C6H4Cl2
InChi Key: OCJBOOLMMGQPQU-RHQRLBAQSA-N
SMILES: [2H]C1=C([2H])C(Cl)=C([2H])C([2H])=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results