accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38565-52-5

FFFFFFFFFFFFFO

CAS RN 38565-52-5

IUPAC Name: 2-(2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptyl)oxirane
Synonyms: 2-(2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptyl)oxirane
Molecular Weight (g/mol): 376.12
Molecular Formula: C9H5F13O
InChi Key: KGYUZRBIQCDOCN-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results