accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3857-25-8

IUPAC Name: (5-methylfuran-2-yl)methanol
Synonyms: (5-methylfuran-2-yl)methanol
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: VOZFDEJGHQWZHU-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)O1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results