accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3858-78-4

IUPAC Name: butan-1-aminium chloride
Synonyms: butan-1-aminium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: ICXXXLGATNSZAV-UHFFFAOYSA-N
SMILES: [Cl-].CCCC[NH3+]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results