accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 3859-41-4

IUPAC Name: cyclopentane-1,3-dione
Synonyms: 1,3-cyclopentanedione
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: LOGSONSNCYTHPS-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)