accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38603-09-7

IUPAC Name: 1,3-dibromo-2-methoxybenzene
Synonyms: 1,3-dibromo-2-methoxybenzene
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: BMZVDHQOAJUZJL-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results