accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHOOHHO

CAS RN 38628-65-8

IUPAC Name: 2-hydroxybut-3-ynoic acid
Synonyms: 2-hydroxybut-3-ynoic acid
Molecular Weight (g/mol): 100.07
Molecular Formula: C4H4O3
InChi Key: LWNHDEQKHFRYMD-UHFFFAOYNA-N
SMILES: OC(C#C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results