accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FF

CAS RN 3863-11-4

IUPAC Name: 3,4-difluoroaniline
Synonyms: benzenamine, 3,4-difluoro-
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: AXNUZKSSQHTNPZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results