accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38641-82-6

IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol
Synonyms: edetic acid; tris buffer
Molecular Weight (g/mol): 413.38
Molecular Formula: C14H27N3O11
InChi Key: VLEIUWBSEKKKFX-UHFFFAOYSA-N
SMILES: NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Grade 
  • (1)
Quantity 
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)