CAS RN 3866-79-3
IUPAC Name:
1-methyl-4-[({3-[(4-methylbenzenesulfonyl)sulfanyl]propyl}sulfanyl)sulfonyl]benzene
Synonyms:
1-methyl-4-({3-[(4-methylbenzenesulfonyl)sulfanyl]propyl}sulfanylsulfonyl)benzene
Molecular Weight (g/mol):
416.58
Molecular Formula:
C17H20O4S4
InChi Key:
QOICHLMIPNOKLN-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)SCCCSS(=O)(=O)C1=CC=C(C)C=C1