accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 386761-71-3

IUPAC Name: 2,4-di-tert-butyl-6-{[(3-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-2,3-dimethylbutan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one; chlororutheniumylium; oxoazanide
Synonyms: 2,4-di-tert-butyl-6-{[(3-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-2,3-dimethylbutan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one; chlororutheniumylium; hyponitrite
Molecular Weight (g/mol): 715.38
Molecular Formula: C36H56ClN3O3Ru
InChi Key: ZQSCHIZFMGABNG-UHFFFAOYSA-M
SMILES: Cl[Ru+].[N-]=O.CC(C)(C)C1=CC(=CNC(C)(C)C(C)(C)NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results