accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 38690-76-5

IUPAC Name: 4-cyanophenyl 4-heptylbenzoate
Synonyms: 4-cyanophenyl 4-heptylbenzoate
Molecular Weight (g/mol): 321.42
Molecular Formula: C21H23NO2
InChi Key: ZWAUTNYPKHVAEZ-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results