CAS RN 38713-56-3
IUPAC Name:
nonyl 4-hydroxybenzoate
Synonyms:
nonyl 4-hydroxybenzoate
Molecular Weight (g/mol):
264.37
Molecular Formula:
C16H24O3
InChi Key:
MBNSKHJDYXPONL-UHFFFAOYSA-N
SMILES:
CCCCCCCCCOC(=O)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
Nonyl 4-Hydroxybenzoate 98.0+%, TCI America™
CAS: 38713-56-3 Molecular Formula: C16H24O3 Molecular Weight (g/mol): 264.365 MDL Number: MFCD00016483 InChI Key: MBNSKHJDYXPONL-UHFFFAOYSA-N Synonym: 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben PubChem CID: 586708 IUPAC Name: nonyl 4-hydroxybenzoate SMILES: CCCCCCCCCOC(=O)C1=CC=C(C=C1)O