accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOCl

CAS RN 3874-54-2

IUPAC Name: 4-chloro-1-(4-fluorophenyl)butan-1-one
Synonyms: 4-chloro-1-(4-fluorophenyl)butan-1-one
Molecular Weight (g/mol): 200.64
Molecular Formula: C10H10ClFO
InChi Key: HXAOUYGZEOZTJO-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCCl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results