accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3875-78-3

IUPAC Name: 6-bromo-3,4-dihydro-2H-1-benzopyran
Synonyms: 6-bromo-3,4-dihydro-2H-1-benzopyran
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: KPFDABVKWKOIME-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results