accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38762-41-3

IUPAC Name: 3-bromo-5-chloroaniline
Synonyms: 3-bromo-5-chloroaniline
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: YYBRLVWWPRAQDX-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)