accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSClOO

CAS RN 38775-38-1

IUPAC Name: hexadecane-1-sulfonyl chloride
Synonyms: hexadecane-1-sulfonyl chloride
Molecular Weight (g/mol): 324.95
Molecular Formula: C16H33ClO2S
InChi Key: YOSVFFVBSPQTTP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCS(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)