accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3878-24-8

IUPAC Name: 6-methyl-2-(thiophen-2-yl)-1H-1,3-benzodiazole
Synonyms: 5-methyl-2-(thiophen-2-yl)-3H-1,3-benzodiazole
Molecular Weight (g/mol): 214.29
Molecular Formula: C12H10N2S
InChi Key: QIYVOSKFSQXPCG-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(NC2=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results