accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3878-55-5

IUPAC Name: 4-methoxy-4-oxobutanoic acid
Synonyms: butanedioic acid, monomethyl ester
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: JDRMYOQETPMYQX-UHFFFAOYSA-N
SMILES: COC(=O)CCC(O)=O
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)