accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SiH3CCH3OBrOSiCH3H3CCH3CH3H3C

CAS RN 387834-41-5

IUPAC Name: {[3-(bromomethylidene)-5-[(tert-butyldimethylsilyl)oxy]cyclohexyl]oxy}(tert-butyl)dimethylsilane
Synonyms: {[3-(bromomethylidene)-5-[(tert-butyldimethylsilyl)oxy]cyclohexyl]oxy}(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 435.59
Molecular Formula: C19H39BrO2Si2
InChi Key: GCYQQWTZQGQJBW-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OC1CC(CC(C1)=CBr)O[Si](C)(C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results