accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 388-82-9

IUPAC Name: 2,2'-difluoro-1,1'-biphenyl
Synonyms: 2,2'-difluoro-1,1'-biphenyl
Molecular Weight (g/mol): 190.19
Molecular Formula: C12H8F2
InChi Key: PXFIPIAXFGAEMJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results