accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38800-20-3

IUPAC Name: 2,3-diphenyl-5-(thiophen-2-yl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 2,3-diphenyl-5-(thiophen-2-yl)-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 340.83
Molecular Formula: C17H13ClN4S
InChi Key: RLHVMOZOYHDIGV-UHFFFAOYSA-M
SMILES: [Cl-].S1C=CC=C1C1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)