accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 388071-27-0

IUPAC Name: tert-butyl N-[2-(3,5-difluorophenyl)-1-(oxiran-2-yl)ethyl]carbamate
Synonyms: tert-butyl N-[2-(3,5-difluorophenyl)-1-(oxiran-2-yl)ethyl]carbamate
Molecular Weight (g/mol): 299.32
Molecular Formula: C15H19F2NO3
InChi Key: NKGKCDXMOMAORK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC(F)=CC(F)=C1)C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results