CAS RN 3883-86-1
IUPAC Name:
2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
Synonyms:
2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
Molecular Weight (g/mol):
298.14
Molecular Formula:
C12H2F8
InChi Key:
QWCHHUZAAGRHDB-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F
1
–
1
of
1
results
1
2,2',3,3',5,5',6,6'-Octafluorobiphenyl 97.0+%, TCI America™
CAS: 3883-86-1 Molecular Formula: C12H2F8 Molecular Weight (g/mol): 298.14 MDL Number: MFCD00000312 InChI Key: QWCHHUZAAGRHDB-UHFFFAOYSA-N Synonym: 4H,4′CH-Octafluorobiphenyl PubChem CID: 77492 IUPAC Name: 2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl SMILES: FC1=CC(F)=C(F)C(=C1F)C1=C(F)C(F)=CC(F)=C1F