CAS RN 3886-69-9
(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
D(+)-alpha-Methylbenzylamine, 99+%, (99% ee)
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.18 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine, 98%
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine, 99+%
CAS: 3886-69-9 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(R)-(+)-1-Phenylethylamine 99.0+%, TCI America™
CAS: 3886-69-9 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064405 InChI Key: RQEUFEKYXDPUSK-SSDOTTSWSA-N Synonym: r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine PubChem CID: 643189 ChEBI: CHEBI:35322 IUPAC Name: (1R)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
Sigma Aldrich L-Fmoc-4-Fluorophenylalanine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich ™-(+)-Alpha-Methylbenzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich ™-(+)-Alpha-Methylbenzylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.