accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38861-88-0

IUPAC Name: 4-(2-methylpropyl)benzoic acid
Synonyms: 4-(2-methylpropyl)benzoic acid
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: VUBBCFWWSKOHTH-UHFFFAOYSA-N
SMILES: CC(C)CC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results