accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONOOH3C(E/Z)

CAS RN 388631-93-4

IUPAC Name: tert-butyl 3-(3-methoxy-3-oxoprop-1-en-1-yl)-1H-indole-1-carboxylate
Synonyms: tert-butyl 3-(3-methoxy-3-oxoprop-1-en-1-yl)indole-1-carboxylate
Molecular Weight (g/mol): 301.34
Molecular Formula: C17H19NO4
InChi Key: VSMVLFKTUMKNRW-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results