accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3887-02-3

IUPAC Name: N,2-dimethylprop-2-enamide
Synonyms: N,2-dimethylprop-2-enamide
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: WFKDPJRCBCBQNT-UHFFFAOYSA-N
SMILES: CNC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results