accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOCH3HOCH3HOCH3HOCH3HONb

CAS RN 38874-17-8

IUPAC Name: pentakis(propan-1-ol) niobium
Synonyms: pentakis(propanol) niobium
Molecular Weight (g/mol): 393.39
Molecular Formula: C15H40NbO5
InChi Key: VXIXBNJIHXAVQS-UHFFFAOYSA-N
SMILES: [Nb].CCCO.CCCO.CCCO.CCCO.CCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results