accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38880-58-9

IUPAC Name: (2-hydroxyethyl)dimethyl(3-sulfopropyl)azanium hydroxide
Synonyms: (2-hydroxyethyl)dimethyl(3-sulfopropyl)azanium hydroxide
Molecular Weight (g/mol): 229.29
Molecular Formula: C7H19NO5S
InChi Key: QDYXZZUKRVDVEQ-UHFFFAOYSA-N
SMILES: [OH-].C[N+](C)(CCO)CCCS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results