accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHOONHOH

CAS RN 38937-66-5

IUPAC Name: N,N'-dihydroxyoctanediamide
Synonyms: N,N'-dihydroxyoctanediamide
Molecular Weight (g/mol): 204.23
Molecular Formula: C8H16N2O4
InChi Key: IDQPVOFTURLJPT-UHFFFAOYSA-N
SMILES: ONC(=O)CCCCCCC(=O)NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results