accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38944-14-8

IUPAC Name: 2-(4-chlorophenyl)pyrrolidine
Synonyms: 2-(4-chlorophenyl)pyrrolidine
Molecular Weight (g/mol): 181.66
Molecular Formula: C10H12ClN
InChi Key: CIHHGGKKRPPWSU-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results