CAS RN 38945-21-0
IUPAC Name:
hydrogen O-(prop-2-en-1-yl)hydroxylamine chloride
Synonyms:
hydrogen O-(prop-2-en-1-yl)hydroxylamine chloride
Molecular Weight (g/mol):
109.55
Molecular Formula:
C3H8ClNO
InChi Key:
XIQUJVRFXPBMHS-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NOCC=C
1
–
1
of
1
results
1
O-Allylhydroxylamine Hydrochloride 98.0+%, TCI America™
CAS: 38945-21-0 Molecular Formula: C3H8ClNO Molecular Weight (g/mol): 109.553 MDL Number: MFCD00012957 InChI Key: XIQUJVRFXPBMHS-UHFFFAOYSA-N PubChem CID: 3084724 IUPAC Name: O-prop-2-enylhydroxylamine;hydrochloride SMILES: C=CCON.Cl