accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38952-62-4

IUPAC Name: 4-(morpholine-4-carbonyl)morpholine
Synonyms: 4-(morpholine-4-carbonyl)morpholine
Molecular Weight (g/mol): 200.24
Molecular Formula: C9H16N2O3
InChi Key: QONGECDDDTYBGS-UHFFFAOYSA-N
SMILES: O=C(N1CCOCC1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results