accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNClNOHH3CH3CCH3

CAS RN 3896-11-5

IUPAC Name: 2-tert-butyl-6-(5-chloro-2H-1,2,3-benzotriazol-2-yl)-4-methylphenol
Synonyms: tinuvin tinuvin-1130 tinuvin-326 bumetrizole tinuvin 326 C17H18clN3O bumetrizol UV absorber-6 bumetrizolum
Molecular Weight (g/mol): 315.8
Molecular Formula: C17H18ClN3O
InChi Key: OCWYEMOEOGEQAN-UHFFFAOYSA-N
SMILES: CC1=CC(N2N=C3C=CC(Cl)=CC3=N2)=C(O)C(=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results