accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 389621-84-5

IUPAC Name: [4-(morpholine-4-carbonyl)phenyl]boronic acid
Synonyms: 4-(morpholine-4-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 235.05
Molecular Formula: C11H14BNO4
InChi Key: KMNLIQJXZPBCDU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)N1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)