accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38980-96-0

IUPAC Name: 4-(trifluoromethyl)benzene-1-carboximidamide hydrochloride
Synonyms: 4-(trifluoromethyl)benzenecarboximidamide hydrochloride
Molecular Weight (g/mol): 224.61
Molecular Formula: C8H8ClF3N2
InChi Key: DKIFMADLURULQV-UHFFFAOYSA-N
SMILES: Cl.NC(=N)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)