accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 389845-43-6

IUPAC Name: [(2,3-dihydro-1-benzofuran-7-yl)methyl](methyl)amine
Synonyms: (2,3-dihydro-1-benzofuran-7-ylmethyl)(methyl)amine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: WVSLYVGYBIMHJY-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results