accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38985-79-4

IUPAC Name: 5-bromo-2-acetamidobenzoate
Synonyms: 5-bromo-2-acetamidobenzoate
Molecular Weight (g/mol): 257.06
Molecular Formula: C9H7BrNO3
InChi Key: QVABAFHRLMDDLM-UHFFFAOYSA-M
SMILES: CC(=O)NC1=CC=C(Br)C=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results