accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNSONOOHBr

CAS RN 389850-21-9

IUPAC Name: 2-(2-imino-2,3,4,5,6,7-hexahydro-1,3-benzothiazol-3-yl)-1-(4-nitrophenyl)ethan-1-one hydrobromide
Synonyms: 2-(2-imino-4,5,6,7-tetrahydro-1,3-benzothiazol-3-yl)-1-(4-nitrophenyl)ethanone hydrobromide
Molecular Weight (g/mol): 398.28
Molecular Formula: C15H16BrN3O3S
InChi Key: NXPCMOGORSWOLH-UHFFFAOYSA-N
SMILES: Br.[O-][N+](=O)C1=CC=C(C=C1)C(=O)CN1C(=N)SC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results