accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39021-62-0

IUPAC Name: 1-methyl-1H-imidazole-5-carbaldehyde
Synonyms: 3-methylimidazole-4-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: BNYKZFOZWZMEJD-UHFFFAOYSA-N
SMILES: CN1C=NC=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results